Merck
  • Spectrofluorimetric quantification of bromazepam using a highly selective optical probe based on Eu3+-bromazepam complex in pharmaceutical and serum samples.
Spectrofluorimetric quantification of bromazepam using a highly selective optical probe based on Eu3+-bromazepam complex in pharmaceutical and serum samples.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2009-09-29)
M S Attia
ABSTRACT
A rapid, simple and sensitive spectrofluorimetric method for determination of trace amount of bromazepam is developed. In phosphate buffer of pH 7.4. The bromazepam enhance the luminescence intensity of the Eu(3+) ion in Eu(3+)-bromazepam complex at lambda(ex)=390nm. The produced luminescence intensity of Eu(3+)-bromazepam complex is in proportion to the concentration of bromazepam. The working range for the determination of bromazepam is 2.3x10(-8) to 6.2x10(-7)M with detection limit (LoD) and quantitative detection limit (LoQ) of 3x10(-9) and 1.2x10(-8)M, respectively. While, the working range, detection limit (LoD) and quantitative detection limit (LoQ) in case of the quantum yield calculations are 3.7x10(-8) to 3.4x10(-7)M with of 3.4x10(-9) and 9.2x10(-8)M, respectively. The enhancement mechanism of the luminescence intensity in the Eu(3+)-bromazepam system has been also explained.
Product Description
Supelco
Bromazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Bromazepam, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.