Merck
  • Increased folate catabolism following total body gamma-irradiation in mice.
Increased folate catabolism following total body gamma-irradiation in mice.
Journal of radiation research (2003-09-19)
Vellappan Kesavan, Madan S Pote, Vipen Batra, Gomathy Viswanathan
ABSTRACT
Concentrations of total folates and their oxidative degradate, para-aminobenzoyl glutamic acid, were determined in mouse after 2-7 Gy total body gamma-irradiation (TBI). Total liver folate levels were drastically reduced by almost 47% over a period of 120 h after TBI with 7 Gy. Oxidative damage, splitting the folate molecule into pterin and p-aminobenzoylglutamic acid (p-ABG), was observed after TBI. p-ABG levels, which 24 h after irradiation were raised by 15%, were further elevated to more than three-fold over the control after 120 h. A dose-dependent increase in the oxidative degradation of folate was observed. The oxidative cleavage of folate may be one factor contributing to folate deficiency in radiation stress.
Product Description
Folic acid impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-(4-Aminobenzoyl)-L-glutamic acid, ≥98% (TLC)