Merck
The use of tailed octamer primers for cycle sequencing.
Nucleic acids research (1998-11-04)
S Ball, M A Reeve, P S Robinson, F Hill, D M Brown, D Loakes
ABSTRACT
Studies have been carried out on the use of octamer oligonucleotides tailed with different base analogues as primers in cycle sequencing reactions. 5-Nitroindole tails improved the performance as primers of a number of octamers. A tail length of three or four 5-nitroindole residues significantly increased the sequencing signal intensity for almost all primers. The use of incomplete libraries of tailed octamer primers for primer walking is discussed.
Sigma-Aldrich
5-Nitroindole, 98%