Merck
  • Radiometric selective inhibition tests for differentiation of Mycobacterium tuberculosis, Mycobacterium bovis, and other mycobacteria.
Radiometric selective inhibition tests for differentiation of Mycobacterium tuberculosis, Mycobacterium bovis, and other mycobacteria.
Journal of clinical microbiology (1985-04-01)
W M Gross, J E Hawkins
ABSTRACT
In the context of a busy reference laboratory, radiometric selective inhibition tests were evaluated for rapid differentiation of Mycobacterium tuberculosis and Mycobacterium bovis and of the M. tuberculosis complex from other mycobacteria. p-Nitro-alpha-acetylamino-beta-hydroxypropiophenone at 5 micrograms and hydroxylamine hydrochloride at 62.5 and 125 micrograms per ml of 7H12 medium were used to separate the M. tuberculosis complex from other mycobacteria (MOTT bacilli). Since it is important epidemiologically to distinguish M. tuberculosis from M. bovis, susceptibility to 1 microgram of thiophene-2-carboxylic acid per ml was also determined radiometrically. By using these three agents as selective inhibitors, M. tuberculosis, M. bovis, and MOTT bacilli were differentiated with a high degree of specificity by a BACTEC radiometric procedure. Results of tests performed on clinical isolates submitted on solid medium to our reference laboratory were available within 5 days.
Sigma-Aldrich
2-Thiophenecarboxylic acid hydrazide, ≥98%
Pricing and availability is not currently available.