Merck
  • Histochemical demonstration of copper and copper-associated protein in the canine liver.
Histochemical demonstration of copper and copper-associated protein in the canine liver.
Veterinary pathology (1985-07-01)
L P Thornburg, M Beissenherz, M Dolan, M F Raisbeck
ABSTRACT
Three different histochemical methods for copper detection were compared. Atomic absorption analysis was used to substantiate the tissue stains. There was good correlation between rhodanine staining and rubeanic acid-stained tissue sections. The orcein reaction for copper-associated protein did not consistently correlate with the methods demonstrating copper. Prolonged staining (72 hours) with rubeanic acid more consistently and clearly detected increased copper in canine livers than did staining with rhodanine. Seventy-two hour staining with rubeanic acid is the method of choice for histochemical detection of copper in canine liver.
Sigma-Aldrich
Dithiooxamide, 97%