Merck
  • Xk-related protein 8 and CED-8 promote phosphatidylserine exposure in apoptotic cells.
Xk-related protein 8 and CED-8 promote phosphatidylserine exposure in apoptotic cells.
Science (New York, N.Y.) (2013-07-13)
Jun Suzuki, Daniel P Denning, Eiichi Imanishi, H Robert Horvitz, Shigekazu Nagata
ABSTRACT
A classic feature of apoptotic cells is the cell-surface exposure of phosphatidylserine (PtdSer) as an "eat me" signal for engulfment. We show that the Xk-family protein Xkr8 mediates PtdSer exposure in response to apoptotic stimuli. Mouse Xkr8(-/-) cells or human cancer cells in which Xkr8 expression was repressed by hypermethylation failed to expose PtdSer during apoptosis and were inefficiently engulfed by phagocytes. Xkr8 was activated directly by caspases and required a caspase-3 cleavage site for its function. CED-8, the only Caenorhabditis elegans Xk-family homolog, also promoted apoptotic PtdSer exposure and cell-corpse engulfment. Thus, Xk-family proteins have evolutionarily conserved roles in promoting the phagocytosis of dying cells by altering the phospholipid distribution in the plasma membrane.
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, SAJ first grade, ≥99.5%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis