Merck
  • Resolution of Clostridium difficile-associated diarrhea in patients with cancer treated with fidaxomicin or vancomycin.
Resolution of Clostridium difficile-associated diarrhea in patients with cancer treated with fidaxomicin or vancomycin.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-05-30)
Oliver A Cornely, Mark A Miller, Bruno Fantin, Kathleen Mullane, Yin Kean, Sherwood Gorbach
ABSTRACT
Patients with cancer are at increased risk for Clostridium difficile-associated diarrhea (CDAD). Little is known about treatment response. Two double-blind trials randomly allocated 1,105 patients with CDAD to fidaxomicin or vancomycin treatment (modified intent-to-treat [mITT]), and 183 of these had cancer. Univariate and multivariate post hoc analyses compared effects of treatment and patient characteristics on cure, recurrence, and sustained response after 4 weeks. Time to resolution of diarrhea (TTROD) was also evaluated. Patients with cancer had a lower cure rate and longer TTROD than patients without cancer. Recurrence rates were similar. Cure was more likely with fidaxomicin than vancomycin (odds ratio [OR] 2.0; P = .065), recurrence was less likely (OR = 0.37; P = .018), and sustained response more frequent (OR = 2.56; P = .003). Under vancomycin, median TTROD was longer in patients with cancer than in those without (123 v 58 hours; log-rank P < .001). With fidaxomicin, median TTROD was not significantly affected by presence of cancer (74 v 54 hours; log-rank P = .145). In the full mITT population, age, hypoalbuminemia, and cancer were inversely associated with clinical cure by multivariate analysis. Study treatment with vancomycin was a significant predictor of recurrence (P < .001). Within the cancer population, low albumin was negatively and fidaxomicin was positively associated with improved cure. For patients with cancer, fidaxomicin treatment was superior to vancomycin, resulting in higher cure and sustained response rates, shorter TTROD, and fewer recurrences.
Product Description
Millipore
Vancomycin supplement, suitable for microbiology
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, meets USP testing specifications
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)