Merck
  • Guidelines for diagnosis, treatment, and prevention of Clostridium difficile infections.
Guidelines for diagnosis, treatment, and prevention of Clostridium difficile infections.
The American journal of gastroenterology (2013-02-27)
Christina M Surawicz, Lawrence J Brandt, David G Binion, Ashwin N Ananthakrishnan, Scott R Curry, Peter H Gilligan, Lynne V McFarland, Mark Mellow, Brian S Zuckerbraun
ABSTRACT
Clostridium difficile infection (CDI) is a leading cause of hospital-associated gastrointestinal illness and places a high burden on our health-care system. Patients with CDI typically have extended lengths-of-stay in hospitals, and CDI is a frequent cause of large hospital outbreaks of disease. This guideline provides recommendations for the diagnosis and management of patients with CDI as well as for the prevention and control of outbreaks while supplementing previously published guidelines. New molecular diagnostic stool tests will likely replace current enzyme immunoassay tests. We suggest treatment of patients be stratified depending on whether they have mild-to-moderate, severe, or complicated disease. Therapy with metronidazole remains the choice for mild-to-moderate disease but may not be adequate for patients with severe or complicated disease. We propose a classification of disease severity to guide therapy that is useful for clinicians. We review current treatment options for patients with recurrent CDI and recommendations for the control and prevention of outbreaks of CDI.
Product Description
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Metronidazole, SAJ first grade, ≥99.0%
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material