Merck
  • Reduction of Fe(III)EDTA by Klebsiella sp. strain FD-3 in NOx scrubber solutions.
Reduction of Fe(III)EDTA by Klebsiella sp. strain FD-3 in NOx scrubber solutions.
Bioresource technology (2013-02-16)
Zuoming Zhou, Guohua Jing, Xiangjiao Zheng
ABSTRACT
Biological reduction of Fe(III) to Fe(II) is a key step in nitrogen oxides (NOx) removal by the integrated chemical absorption-biological reduction method, which determines the concentration of Fe(II) in the scrubbing liquid. A new Fe(III)EDTA reduction strain, named as FD-3, was isolated from mixed cultures used in the integrated NOx removal process and identified as Klebsiella sp. by 16S rDNA sequence analysis. The reduction abilities of FD-3 and the influence of nitrogen-containing compounds (Fe(II)EDTA-NO, NO3(-) and NO2(-)) and sulfur-containing compounds (SO4(2-), SO3(2-)) on the Fe(III)EDTA reduction were investigated. The results indicated that strain FD-3 could reduce Fe(III)EDTA efficiently. NO3(-), NO2(-) and Fe(II)EDTA-NO inhibit the reduction of Fe(III)EDTA and could also serve as electron acceptor for strain FD-3. SO3(2-) inhibited Fe(III)EDTA reduction while SO4(2-) had no obviously effect on Fe(III)EDTA reduction. The relationship between cell growth and Fe(III)EDTA reduction could be described by the models based on Logistic equation.
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, SAJ special grade, ≥95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, BioReagent, suitable for plant cell culture, crystalline