Merck
  • Controlled soft-template synthesis of ultrathin C@FeS nanosheets with high-Li-storage performance.
Controlled soft-template synthesis of ultrathin C@FeS nanosheets with high-Li-storage performance.
ACS nano (2012-05-10)
Chen Xu, Yi Zeng, Xianhong Rui, Ni Xiao, Jixin Zhu, Wenyu Zhang, Jing Chen, Weiling Liu, Huiteng Tan, Huey Hoon Hng, Qingyu Yan
ABSTRACT
We report a facile approach to prepare carbon-coated troilite FeS (C@FeS) nanosheets via surfactant-assisted solution-based synthesis. 1-Dodecanethiol is used as both the sulfur source and the surfactant, which may form different-shaped micelles to direct the growth of nanostructures. Under appropriate growth conditions, the iron and sulfur atoms react to form thin layers of FeS while the hydrocarbon tails of 1-dodecanethiol separate the thin FeS layers, which turn to carbon after annealing in Ar. Such an approach can be extended to grow C@FeS nanospheres and nanoplates by modifying the synthesis parameters. The C@FeS nanosheets display excellent Li storage properties with high specific capacities and stable charge/discharge cyclability, especially at fast charge/discharge rates.
Sigma-Aldrich
Iron(II) sulfide, SAJ first grade, ≥50.0%
Sigma-Aldrich
Iron(II) sulfide, −100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) sulfide, technical grade