Merck
  • A phytochemical approach to experimental metabolic syndrome-associated renal damage and oxidative stress.
A phytochemical approach to experimental metabolic syndrome-associated renal damage and oxidative stress.
Rejuvenation research (2012-04-27)
Francesco Marotta, Archana Kumari, Roberto Catanzaro, Umberto Solimene, Shalini Jain, Emilio Minelli, Masatoshi Harada
ABSTRACT
The aim of this study was to evaluate the effect of DTS-phytocompound on oxidant-antioxidant balance and protein damage in the kidneys of rats administered high doses of fructose. Adult male Wistar rats were divided into four groups. Group A received a control diet, whereas groups B and C were fed a high-fructose diet (60 g/100 g), the latter with additional DTS (50 mg/kg per day) for 60 days. Lipo- and nitro-peroxidation together with α-smooth muscle actin (α-SMA) expression in the glomerular and interstitial tissue of the kidneys were measured after 60 days. Fructose-fed rats showed significantly higher lipoperoxidation, 2,4-dinitrophenol and 3-nitrotyrosine protein adducts, and upregulation of α-SMA in the kidney. DTS significantly decreased such redox unbalance in renal tissue, while partially downregulating α-SMA (p<0.01). These data suggest the potential clinical benefit of DTS in protecting the kidneys from metabolic syndrome-associated changes; gender-related analysis is under way.
Sigma-Aldrich
2,4-Dinitrophenol, SAJ special grade, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol solution, reference material, 5000 μg/mL in methanol