Merck
Ecotoxicological risks associated with tannery effluent wastewater.
Environmental toxicology and pharmacology (2012-04-24)
Lubna Shakir, Sohail Ejaz, Muhammad Ashraf, Naureen Aziz Qureshi, Aftab Ahmad Anjum, Imran Iltaf, Aqeel Javeed
ABSTRACT
The problem of water pollution acquires greater relevance in the context of a developing agrarian economy like Pakistan. Even though, the leather industry is a leading economic sector in Pakistan, there is an increasing environmental concern regarding tanneries because they produce large amounts of potentially toxic wastewater containing both trivalent and hexavalent chromium, which are equally hazardous for human population, aquaculture and agricultural activities in the area. Therefore, we defined the scope of the present study as to employ different bioassays to determine the eco-toxic potential of tannery effluent wastewater (TW) and its chromium based components, i.e., potassium dichromate (K(2)Cr(2)O(7)) and chromium sulfate Cr(2)(SO(4))(3). Particle-induced X-ray emission (PIXE) analysis of TW was carried out to determine the concentration of chromium in TW and then equal concentrations of hexavalent (K(2)Cr(2)O(7)) and trivalent chromium Cr(2)(SO(4))(3) were obtained for this study. Cytotoxicity assay, artemia bioassay and phytotoxicity assay was utilized to investigate the eco-toxicological potential of different concentrations of TW, K(2)Cr(2)O(7) and Cr(2)(SO(4))(3). All the dilutions of TW, K(2)Cr(2)O(7) and Cr(2)(SO(4))(3) presented concentration dependent cytotoxic effects in these assays. The data clearly represents that among all three tested materials, different dilutions of K(2)Cr(2)O(7) caused significantly more damage (P<0.001) to vero cell, brine shrimp and germination of maize seeds. Interestingly, the overall toxicity effects of TW treated groups were subsequent to K(2)Cr(2)O(7) treated group. Based on biological evidences presented in this article, it is concluded that hexavalent chromium (K(2)Cr(2)O(7)) and TW has got significant eco-damaging potential clearly elaborating that environmental burden in district Kasur is numerous and high levels of chromium is posing a considerable risk to the human population, aquaculture and agricultural industry that can obliterate ecosystem surrounding the tanneries.
Sigma-Aldrich
Chromium standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Cr, 1000 ppm Cr
Sigma-Aldrich
Potassium dichromate solution, 1/60 M
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Supelco
Potassium dichromate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%