Merck
  • Anti-inflammatory effects of arbutin in lipopolysaccharide-stimulated BV2 microglial cells.
Anti-inflammatory effects of arbutin in lipopolysaccharide-stimulated BV2 microglial cells.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2012-04-11)
Hyo-Jong Lee, Kyu-Won Kim
ABSTRACT
Arbutin, which is found in the genus Arctostaphylos, is an anti-oxidant and a depigmenting agent. The present study was designed to validate the anti-inflammatory effect of arbutin. The anti-inflammatory properties of arbutin were studied using a lipopolysaccharide (LPS)-stimulated murine BV2 microglial cells model. As inflammatory parameters, the production of nitric oxide (NO), inducible NO synthase (iNOS), cyclooxygenase-2 (COX-2), tumor necrosis factor-α (TNF-α), interleukin-1β (IL-1β), monocyte chemoattractant protein-1 (MCP-1), and interleukin-6 (IL-6) were evaluated. We also examined the expression of ninjurin1 (Ninj1) and the adhesion activity of BV2 cells. Finally, we analyzed the activation of the nuclear factor-κB (NF-κB) signaling pathway. Arbutin suppressed LPS-induced production of NO and expression of iNOS and COX-2 in a dose-dependent manner without causing cellular toxicity. Arbutin also significantly reduced generation of proinflammatory cytokines, including IL-1β and TNF-α, and other inflammation-related genes such as MCP-1 and IL-6. Additionally, arbutin suppressed the adhesion activity of BV2 cells and the expression of an important adhesion molecule, Ninj1, in LPS-stimulated murine BV2 cells. Furthermore, arbutin inhibited nuclear translocation and the transcriptional activity of NF-κB. Taken together, our results suggest that arbutin might be useful for treating the inflammatory and deleterious effects of BV2 microglial cells activation in response to LPS stimulation.
Product Description
Supelco
Arbutin, analytical standard
Sigma-Aldrich
Arbutin, ≥98% (HPLC)
Arbutin, European Pharmacopoeia (EP) Reference Standard