Merck
  • Zinc PVA versus potassium dichromate for preservation of microsporidian spores of human origin.
Zinc PVA versus potassium dichromate for preservation of microsporidian spores of human origin.
Parasitology research (2012-03-20)
Hend Aly El-Taweel, Mona Mohammed Tolba, Hayam Abdelmonem Sadaka, Lobna Abdelaziz El-Zawawy, Mervat Mostafa Osman
ABSTRACT
Microsporidia are emerging opportunistic parasites. Preservation of the biological properties of microsporidian spores is often required in research work. The present study compared two preservatives; zinc polyvinyl alcohol (zinc PVA) and potassium dichromate solutions for preservation of microsporidian spores separated from human faecal samples. After 0, 1, 2 and 4 months of storage, morphological features and staining characters of the spores were assessed by light microscopy in modified trichrome-stained smears and their viability percentages were calculated using acridine orange/ethidium bromide mixture. Also, spore infectivity was evaluated by faecal spore shedding and intestinal spore load in mice orally inoculated with the preserved spores. Results revealed that morphological features, staining characters and viability of the spores were maintained in both solutions throughout the study period. Spore infectivity was completely preserved in zinc PVA solution but showed significant reduction in potassium dichromate solution at the fourth month of the preservation duration.
Sigma-Aldrich
Chromium standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Cr, 1000 ppm Cr
Sigma-Aldrich
Potassium dichromate solution, 1/60 M
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Supelco
Potassium dichromate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%