Merck
  • Light-into-heat conversion in La2O3:Er(3+)-Yb3+ phosphor: an incandescent emission.
Light-into-heat conversion in La2O3:Er(3+)-Yb3+ phosphor: an incandescent emission.
Optics letters (2012-03-02)
A K Singh, S Singh, D Kumar, D K Rai, S B Rai, K Kumar
ABSTRACT
Low-power-threshold cw laser-induced incandescence (CWLII) has been observed in La(2)O(3):Er(3+)-Yb(3+) phosphor on excitation by a 976 nm IR laser. It is suggested that incandescence originates from the extensive heating induced by the nonradiative processes taking place following the laser excitation. Other mechanisms for similar observations have also been suggested in the literature and have been discussed with the present observations. The estimated temperature for the CWLII approaches around 2650 K, and this seems to provide an effective way to rapidly attain high temperature in nano/microvolumes of phosphor. The phosphor exhibited efficient upconversion, and the ratio of the (2)H(11/2)→(4)I(15/2) and (4)S(3/2)→(4)I(15/2) band intensities of Er(3+) permits measurement of the temperature rise, from a distance.
Product Description
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis