Merck
  • Ameliorating effect of selenium on chromium (VI)-induced oxidative damage in the brain of adult rats.
Ameliorating effect of selenium on chromium (VI)-induced oxidative damage in the brain of adult rats.
Journal of physiology and biochemistry (2012-02-22)
Nejla Soudani, Afef Troudi, Ibtissem Ben Amara, Hanen Bouaziz, Tahia Boudawara, Najiba Zeghal
ABSTRACT
Chromium is known for its wide toxic manifestations. This experiment aims to evaluate the effect of selenium against oxidative stress induced by chromium in the cerebrum and cerebellum. Female Wistar rats were randomly divided into four groups of six each: group I served as controls which received the standard diet; group II received drinking water K(2)Cr(2)O(7) alone (700 ppm); group III received both K(2)Cr(2)O(7) and Se (0.5 mg Na(2)SeO(3)/kg of diet); and group IV received Se (0.5 mg/kg of diet) for 3 weeks. The exposure of rats to K(2)Cr(2)O(7) promoted oxidative stress in the cerebrum and cerebellum with an increase in malondialdehyde and a decrease of nonenzymatic antioxidant levels such as glutathione, nonprotein thiol, and vitamin C. An increase of enzyme activities like catalase, glutathione peroxidase, and superoxide dismutase activities was also observed. Acetylcholinesterase activity was inhibited after treatment with K(2)Cr(2)O(7). Co-administration of Se restored the parameters cited above. The histopathological findings confirmed the biochemical results.
Sigma-Aldrich
Chromium standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Cr, 1000 ppm Cr
Sigma-Aldrich
Potassium dichromate solution, 1/60 M
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Supelco
Potassium dichromate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%