Merck
  • Alteration of mitochondrial efficiency affects oxidative balance, development and growth in frog (Rana temporaria) tadpoles.
Alteration of mitochondrial efficiency affects oxidative balance, development and growth in frog (Rana temporaria) tadpoles.
The Journal of experimental biology (2012-02-11)
Karine Salin, Emilien Luquet, Benjamin Rey, Damien Roussel, Yann Voituron
ABSTRACT
Mitochondria are known to play a central role in life history processes, being the main source of reactive oxygen species (ROS), which promote oxidative constraint. Surprisingly, although the main role of the mitochondria is to produce ATP, the plasticity of mitochondrial ATP generation has received little attention in life history studies. Yet, mitochondrial energy transduction represents the physiological link between environmental resources and energy allocated to animal performance. Studying both facets of mitochondrial functioning (ATP and ROS production) would allow better understanding of the proximate mechanisms underlying life history. We have experimentally modulated the mitochondrial capacity to generate ROS and ATP during larval development of Rana temporaria tadpoles, via chronic exposure (34 days) to a mitochondrial uncoupler (2,4-dinitrophenol, dNP). The aim was to better understand the impact of mitochondrial uncoupling on both responses in terms of oxidative balance, energy input (oxygen and feeding consumption) and energy output (growth and development of the tadpole). Exposure to 2,4-dNP reduced mitochondrial ROS generation, total antioxidant defences and oxidative damage in treated tadpoles compared with controls. Despite the beneficial effect of dNP on oxidative status, development and growth rates of treated tadpoles were lower than those in the control group. Treatment of tadpoles with 2,4-dNP promoted a mild mitochondrial uncoupling and enhanced metabolic rate. These tadpoles did not increase their food consumption, and thus failed to compensate for the energy loss elicited by the decrease in the efficiency of ATP production. These data suggest that the cost of ATP production, rather than the oxidative balance, is the parameter that constrains growth/development of tadpoles, highlighting the central role of energy transduction in larval performance.
Sigma-Aldrich
2,4-Dinitrophenol, SAJ special grade, ≥98.0%
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Supelco
2,4-Dinitrophenol solution, reference material, 5000 μg/mL in methanol