Merck
  • Simultaneous detection of NADH and H₂O₂ using flow injection analysis based on a bifunctional poly(thionine)-modified electrode.
Simultaneous detection of NADH and H₂O₂ using flow injection analysis based on a bifunctional poly(thionine)-modified electrode.
Biosensors & bioelectronics (2012-01-24)
Selvaraj Baskar, Jen-Lin Chang, Jyh-Myng Zen
ABSTRACT
We report here a novel detection scheme for simultaneous detection of NADH and H(2)O(2) based on a bifunctional poly(thionine)-modified electrode. Electropolymerization of thionine on a "preanodized" screen-printed carbon electrode effectively lowers the oxidation potential of NADH to 0.15 V (vs. Ag/AgCl). Since poly(thionine) is also a well known electrochemical mediator for H(2)O(2) reduction, we further developed a poly(thionine)-modified ring disk electrode for simultaneous measurement of nicotinamide adenine dinucleotide (NADH) and hydrogen peroxide (H(2)O(2)) by flow injection analysis. By applying the optimized detection potentials of 0.2V and -0.2V at disk and ring electrodes, respectively, this system allows the simultaneous measurement of both analytes with good sensitivity (0.13 μA/mM for H(2)O(2) and 0.34 μA/mM for NADH) and limit of detection (1.74 μM and 26.0 μM for NADH and H(2)O(2)). This opens the possibility of a whole series of biosensor applications.
Sigma-Aldrich
Thionin acetate salt, certified by the BSC, Dye content ≥85 %