Merck
  • Photocurrents at polarized liquid|liquid interfaces enhanced by a gold nanoparticle film.
Photocurrents at polarized liquid|liquid interfaces enhanced by a gold nanoparticle film.
Physical chemistry chemical physics : PCCP (2011-09-13)
Delphine Schaming, Mohamad Hojeij, Nathalie Younan, Hirohisa Nagatani, Hye Jin Lee, Hubert H Girault
ABSTRACT
Photocurrent responses associated with the interfacial quenching of the photo-excited water-soluble zinc meso-tetra(4-carboxyphenyl)porphyrin (ZnTPPC) by ferrocene have been studied at a water|1,2-dichloroethane interface in the absence and in the presence of adsorbed gold nanoparticles. Upon addition of methanol, a mirror-like gold film is formed and an important enhancement of the photocurrent response can be observed. Intensity modulated photocurrent spectroscopy experiments (IMPS) have been performed, in order to deconvolute in the frequency domain the contribution from the competition between the recombination and the product separation arising after the electron transfer, and the attenuation associated with the resistance and interfacial capacitance (RC(int)) time constant of the cell.
Product Description
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, JIS special grade, ≥99.5%
Supelco
1,2–Dichloroethane solution, certified reference material, 200 μg/mL in methanol
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%