Merck
  • Isatin (indole-2,3-dione) in urine and tissues. Detection and determination by gas chromatography-mass spectrometry.
Isatin (indole-2,3-dione) in urine and tissues. Detection and determination by gas chromatography-mass spectrometry.
Journal of chromatography (1991-01-02)
J M Halket, P J Watkins, A Przyborowska, B L Goodwin, A Clow, V Glover, M Sandler
ABSTRACT
A simple procedure based upon capillary column gas chromatography-mass spectrometry (GC-MS) is described for the detection and determination of isatin (indole-2,3-dione) in body fluids and tissues. After addition of 5-methylisatin as internal standard to urine or tissue homogenates, organic extracts are dried and derivatized successively with hydroxylamine hydrochloride and the reagent N-tert.-butyldimethylsilyl-N-methyltrifluoroacetamide (MTBSTFA). The tert.-butyldimethylsilyl derivatives obtained show good GC-MS properties and allow quantification by selected-ion monitoring of m/z 333 (isatin) and m/z 347 (internal standard). Adult and newborn human urine output values lie in the ranges 0.4-3.2 mg/mmol of creatinine (5-30 mg per 24 h) and 0.002-0.518 mg/mmol of creatinine, respectively. There is a discontinuous regional distribution in rat tissues. The GC-MS properties of a number of derivatives formed by successive reaction of isatin with hydroxylamine hydrochloride (or methoxyaminehydrochloride or ethoxyamine hydrochloride) and MTBSTFA, bis(trimethylsiyl)trifluoroacetamide, pentafluoropropionic anhydride or pentafluorobenzyl bromide are also described.
Sigma-Aldrich
Pentafluoropropionic anhydride, 99%
Supelco
Pentafluoropropionic anhydride, for GC derivatization, LiChropur, 99%
Sigma-Aldrich
5-Methylisatin, 95%
Sigma-Aldrich
Pentafluoropropionic anhydride, purum, ≥97.0% (GC)