Merck
  • Thioredoxin targets of the plant chloroplast lumen and their implications for plastid function.
Thioredoxin targets of the plant chloroplast lumen and their implications for plastid function.
Proteomics (2010-01-06)
Michael Hall, Alejandro Mata-Cabana, Hans-Erik Akerlund, Francisco J Florencio, Wolfgang P Schröder, Marika Lindahl, Thomas Kieselbach
ABSTRACT
The light-dependent regulation of stromal enzymes by thioredoxin (Trx)-catalysed disulphide/dithiol exchange is known as a classical mechanism for control of chloroplast metabolism. Recent proteome studies show that Trx targets are present not only in the stroma but in all chloroplast compartments, from the envelope to the thylakoid lumen. Trx-mediated redox control appears to be a common feature of important pathways, such as the Calvin cycle, starch synthesis and tetrapyrrole biosynthesis. However, the extent of thiol-dependent redox regulation in the thylakoid lumen has not been previously systematically explored. In this study, we addressed Trx-linked redox control in the chloroplast lumen of Arabidopsis thaliana. Using complementary proteomics approaches, we identified 19 Trx target proteins, thus covering more than 40% of the currently known lumenal chloroplast proteome. We show that the redox state of thiols is decisive for degradation of the extrinsic PsbO1 and PsbO2 subunits of photosystem II. Moreover, disulphide reduction inhibits activity of the xanthophyll cycle enzyme violaxanthin de-epoxidase, which participates in thermal dissipation of excess absorbed light. Our results indicate that redox-controlled reactions in the chloroplast lumen play essential roles in the function of photosystem II and the regulation of adaptation to light intensity.
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)
Sigma-Aldrich
Bromobimane, suitable for fluorescence, BioReagent, ≥95% (HPCE)