Merck
  • Crystallization and preliminary crystallographic analysis of a plant type III polyketide synthase that produces benzalacetone.
Crystallization and preliminary crystallographic analysis of a plant type III polyketide synthase that produces benzalacetone.
Acta crystallographica. Section F, Structural biology and crystallization communications (2008-04-09)
Hiroyuki Morita, Michikazu Tanio, Shin Kondo, Ryohei Kato, Kiyofumi Wanibuchi, Hiroshi Noguchi, Shigetoshi Sugio, Ikuro Abe, Toshiyuki Kohno
ABSTRACT
Benzalacetone synthase (BAS) from Rheum palmatum is a plant-specific type III polyketide synthase that catalyzes the one-step decarboxylative condensation of 4-coumaroyl-CoA with malonyl-CoA to produce the diketide 4-(4-hydroxyphenyl)-but-3-en-2-one. Recombinant BAS expressed in Escherichia coli was crystallized by the sitting-drop vapour-diffusion method. The crystals belong to space group P2(1), with unit-cell parameters a = 54.6, b = 89.6, c = 81.1 A, alpha = gamma = 90.0, beta = 100.5 degrees . Diffraction data were collected to 1.8 A resolution using synchrotron radiation at BL24XU of SPring-8.
Sigma-Aldrich
trans-4-Phenyl-3-buten-2-one, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Phenyl-3-buten-2-one, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzylideneacetone, ≥98%, FG
Pricing and availability is not currently available.