Merck
  • Lithium diisopropylamide-mediated enolization: catalysis by hemilabile ligands.
Lithium diisopropylamide-mediated enolization: catalysis by hemilabile ligands.
Journal of the American Chemical Society (2006-08-03)
Antonio Ramirez, Xiufeng Sun, David B Collum
ABSTRACT
Structural, kinetic, and computational studies reveal the mechanistic complexities of a lithium diisopropylamide (LDA)-mediated ester enolization. Hemilabile amino ether MeOCH2CH2NMe2, binding as an eta1 (ether-bound) ligand in the reactant and as an eta2 (chelating) ligand in the transition structure, accelerates the enolization 10,000-fold compared with n-BuOMe. At the onset of the reaction, a dimer-based enolization prevails. As the reaction proceeds, significantly less reactive LDA-enolate mixed dimers appear and divert the reaction through monomer- and mixed dimer-based pathways. The mechanistic and computational investigations lead to a proof-of-principle ligand-catalyzed enolization in which an ancillary ligand allows the catalytic ligand to re-enter the catalytic cycle.
Sigma-Aldrich
Lithium diisopropylamide, 97%
Sigma-Aldrich
Diisopropylamine, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Lithium diisopropylamide solution, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Diisopropylamine, purified by redistillation, 99.95%
Sigma-Aldrich
Diisopropylamine, ≥99.5%
Supelco
Diisopropylamine, analytical standard
Sigma-Aldrich
Diisopropylamine, SAJ first grade, ≥98.0%