Merck
Cap1p attenuates the apoptosis of Candida albicans.
The FEBS journal (2013-03-23)
Bao-Di Dai, Yan Wang, Lan-Xue Zhao, De-Dong Li, Ming-Bang Li, Yong-Bing Cao, Yuan-Ying Jiang
ABSTRACT
Candida albicans is the most common opportunistic fungal pathogen and its apoptosis is inducible by environmental stress. Based on our previous finding that transcription factor Cap1p was involved in baicalein-induced apoptosis, the present study aimed to further clarify the role of Cap1p in apoptosis by observing the impact of CAP1 deletion on cell fate. It was found that apoptotic stimulation with amphotericin B, acetic acid and hydrogen peroxide increased the number of apoptotic and necrotic cells, caspase activity and the accumulation of reactive oxygen species, whereas it decreased the mitochondrial membrane potential and intracellular ATP level in the cap1Δ/Δ mutant. The cell fate was, at least partly, caused by glutathione depletion and attenuation of the expression of the glutathione reductase gene in the cap1Δ/Δ mutant. Collectively, our data suggest that Cap1p participated in the apoptosis of C. albicans by regulating the expression of the glutathione reductase gene and glutathione content.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Glutathione Reductase human, buffered aqueous solution, ≥10 units/mg protein, recombinant, expressed in E. coli
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard