Merck
  • DNA Methylation in Recurrent Glioblastomas: Increased TEM8 Expression Activates the Src/PI3K/AKT/GSK-3β/B-Catenin Pathway.
DNA Methylation in Recurrent Glioblastomas: Increased TEM8 Expression Activates the Src/PI3K/AKT/GSK-3β/B-Catenin Pathway.
Cancer genomics & proteomics (2024-08-28)
Paramita Kundu, Ruchi Jain, Nandaki Nag Kanuri, Arivazhagan Arimappamagan, Vani Santosh, Paturu Kondaiah
ABSTRACT
Glioblastomas (GBM) are infiltrative malignant brain tumors which mostly recur within a year's time following surgical resection and chemo-radiation therapy. Studies on glioblastoma cells following radio-chemotherapy, have been demonstrated to induce trans-differentiation, cellular plasticity, activation of DNA damage response and stemness. As glioblastomas are heterogenous tumors that develop treatment resistance and plasticity, we investigated if there exist genome-wide DNA methylation changes in recurrent tumors. Utilizing genome-wide DNA methylation arrays, we compared the DNA methylation profile of 11 primary (first occurrence) tumors with 13 recurrent (relapsed) GBM, to delineate the contribution of epigenetic changes associated with therapy exposure, therapy resistance, and relapse of disease. Our data revealed 1,224 hypermethylated- and 526 hypomethylated-probes in recurrent glioblastomas compared to primary disease. We found differential methylation of solute carrier and ion channel genes, interleukin receptor/ligand genes, tumor-suppressor genes and genes associated with metastasis. We functionally characterized one such hypomethylated-up-regulated gene, namely anthrax toxin receptor 1/tumor endothelial marker 8 (ANTXR1/TEM8), whose expression was validated to be significantly up-regulated in recurrent glioblastomas compared to primary tumors and confirmed by immunohistochemistry. Using overexpression and knockdown approaches, we showed that TEM8 induces proliferation, invasion, migration, and chemo-radioresistance in glioblastoma cells. Additionally, we demonstrated a novel mechanism of β-catenin stabilization and activation of the β-catenin transcriptional program due to TEM8 overexpression via a Src/PI3K/AKT/GSK3β/β-catenin pathway. We report genome-wide DNA methylation changes in recurrent GBM and suggest involvement of the TEM8 gene in GBM recurrence and progression.
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-phospho-ILK (Ser246) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Focal Adhesion Kinase Inhibitor I, The Focal Adhesion Kinase Inhibitor I, also referenced under CAS 4506-66-5, controls the biological activity of Foca Adhesion Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.