Merck
  • Sialyltransferase ST3Gal-IV controls CXCR2-mediated firm leukocyte arrest during inflammation.
Sialyltransferase ST3Gal-IV controls CXCR2-mediated firm leukocyte arrest during inflammation.
The Journal of experimental medicine (2008-06-04)
David Frommhold, Andreas Ludwig, M Gabriele Bixel, Alexander Zarbock, Inna Babushkina, Melitta Weissinger, Sandra Cauwenberghs, Lesley G Ellies, Jamey D Marth, Annette G Beck-Sickinger, Michael Sixt, Bärbel Lange-Sperandio, Alma Zernecke, Ernst Brandt, Christian Weber, Dietmar Vestweber, Klaus Ley, Markus Sperandio
ABSTRACT
Recent in vitro studies have suggested a role for sialylation in chemokine receptor binding to its ligand (Bannert, N., S. Craig, M. Farzan, D. Sogah, N.V. Santo, H. Choe, and J. Sodroski. 2001. J. Exp. Med. 194:1661-1673). This prompted us to investigate chemokine-induced leukocyte adhesion in inflamed cremaster muscle venules of alpha2,3 sialyltransferase (ST3Gal-IV)-deficient mice. We found a marked reduction in leukocyte adhesion to inflamed microvessels upon injection of the CXCR2 ligands CXCL1 (keratinocyte-derived chemokine) or CXCL8 (interleukin 8). In addition, extravasation of ST3Gal-IV(-/-) neutrophils into thioglycollate-pretreated peritoneal cavities was significantly decreased. In vitro assays revealed that CXCL8 binding to isolated ST3Gal-IV(-/-) neutrophils was markedly impaired. Furthermore, CXCL1-mediated adhesion of ST3Gal-IV(-/-) leukocytes at physiological flow conditions, as well as transendothelial migration of ST3Gal-IV(-/-) leukocytes in response to CXCL1, was significantly reduced. In human neutrophils, enzymatic desialylation decreased binding of CXCR2 ligands to the neutrophil surface and diminished neutrophil degranulation in response to these chemokines. In addition, binding of alpha2,3-linked sialic acid-specific Maackia amurensis lectin II to purified CXCR2 from neuraminidase-treated CXCR2-transfected HEK293 cells was markedly impaired. Collectively, we provide substantial evidence that sialylation by ST3Gal-IV significantly contributes to CXCR2-mediated leukocyte adhesion during inflammation in vivo.
Product Description
Millipore
Thioglycollate medium, Microbiologically tested, NutriSelect® Plus
Pricing and availability is not currently available.