Merck
  • Modulation of 5-HT release by dynorphin mediates social deficits during opioid withdrawal.
Modulation of 5-HT release by dynorphin mediates social deficits during opioid withdrawal.
Neuron (2022-10-07)
Matthew B Pomrenze, Daniel F Cardozo Pinto, Peter A Neumann, Pierre Llorach, Jason M Tucciarone, Wade Morishita, Neir Eshel, Boris D Heifets, Robert C Malenka
ABSTRACT
Social isolation during opioid withdrawal is a major contributor to the current opioid addiction crisis. We find that sociability deficits during protracted opioid withdrawal in mice require activation of kappa opioid receptors (KORs) in the nucleus accumbens (NAc) medial shell. Blockade of release from dynorphin (Pdyn)-expressing dorsal raphe neurons (DRPdyn), but not from NAcPdyn neurons, prevents these deficits in prosocial behaviors. Conversely, optogenetic activation of DRPdyn neurons reproduced NAc KOR-dependent decreases in sociability. Deletion of KORs from serotonin (5-HT) neurons, but not from NAc neurons or dopamine (DA) neurons, prevented sociability deficits during withdrawal. Finally, measurements with the genetically encoded GRAB5-HT sensor revealed that during withdrawal KORs block the NAc 5-HT release that normally occurs during social interactions. These results define a neuromodulatory mechanism that is engaged during protracted opioid withdrawal to induce maladaptive deficits in prosocial behaviors, which in humans contribute to relapse.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-mCherry Antibody, clone 1C51, clone 1C51, from mouse