Merck
  • ATR-binding lncRNA ScaRNA2 promotes cancer resistance through facilitating efficient DNA end resection during homologous recombination repair.
ATR-binding lncRNA ScaRNA2 promotes cancer resistance through facilitating efficient DNA end resection during homologous recombination repair.
Journal of experimental & clinical cancer research : CR (2023-09-30)
Yuanyuan Chen, Hui Shen, Tingting Liu, Kun Cao, Zhijie Wan, Zhipeng Du, Hang Wang, Yue Yu, Shengzhe Ma, Edward Lu, Wei Zhang, Jianming Cai, Fu Gao, Yanyong Yang
ABSTRACT
Our previous study first showed that ATR-binding long noncoding RNA (lncRNA) is necessary for ATR function and promotes cancer resistance. However, the specific lncRNAs instrumental in ATR activation remain largely unclear, which limits our comprehensive understanding of this critical biological process. RNA immunoprecipitation (RIP) followed by RNA sequencing was employed to identify ATR-binding lncRNAs, which were further validated using RIP-qPCR assays. Immunofluorescence staining and Western blotting were applied to detect the activation of DNA damage repair factors. After the effect of scaRNA2 on cellular sensitivity to DNA-damaging reagents was determined, the effects of scaRNA2 on radiotherapy were investigated in patient-derived organoids and xenograft preclinical models. The clinical relevance of scaRNA2 was also validated in tissues isolated from rectal cancer patients. ScaRNA2 was identified as the most enriched ATR-binding lncRNA and was found to be essential for homologous recombination (HR) mediated DNA damage repair. Furthermore, scaRNA2 knockdown abrogated the recruitment of ATR and its substrates in response to DNA damage. Mechanistically, scaRNA2 was observed to be necessary for Exo1-mediated DNA end resection and bridged the MRN complex to ATR activation. Knockdown of scaRNA2 effectively increased the sensitivity of cancer cells to multiple kinds of DNA damage-related chemoradiotherapy. Preclinically, knockdown of scaRNA2 improved the effects of radiotherapy on patient-derived organoids and xenograft models. Finally, an increase in scaRNA2 colocalized with ATR was also found in clinical patients who were resistant to radiotherapy. ScaRNA2 was identified as the most abundant lncRNA bound to ATR and was demonstrated to bridge DNA end resection to ATR activation; thus, it could be applied as a potent target for combined cancer treatments with chemoradiotherapy.
Sigma-Aldrich
Rabbit Anti-Human IgG Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.