Merck
  • Incongruence between transcriptional and vascular pathophysiological cell states.
Incongruence between transcriptional and vascular pathophysiological cell states.
Nature cardiovascular research (2023-09-25)
Macarena Fernández-Chacón, Severin Mühleder, Alvaro Regano, Lourdes Garcia-Ortega, Susana F Rocha, Carlos Torroja, Maria S Sanchez-Muñoz, Mariya Lytvyn, Verónica Casquero-Garcia, Macarena De Andrés-Laguillo, Lars Muhl, Michael M Orlich, Konstantin Gaengel, Emilio Camafeita, Jesús Vázquez, Alberto Benguría, M Luisa Iruela-Arispe, Ana Dopazo, Fátima Sánchez-Cabo, Hannah Carter, Rui Benedito
ABSTRACT
The Notch pathway is a major regulator of endothelial transcriptional specification. Targeting the Notch receptors or Delta-like ligand 4 (Dll4) dysregulates angiogenesis. Here, by analyzing single and compound genetic mutants for all Notch signaling members, we find significant differences in the way ligands and receptors regulate liver vascular homeostasis. Loss of Notch receptors caused endothelial hypermitogenic cell-cycle arrest and senescence. Conversely, Dll4 loss triggered a strong Myc-driven transcriptional switch inducing endothelial proliferation and the tip-cell state. Myc loss suppressed the induction of angiogenesis in the absence of Dll4, without preventing the vascular enlargement and organ pathology. Similarly, inhibition of other pro-angiogenic pathways, including MAPK/ERK and mTOR, had no effect on the vascular expansion induced by Dll4 loss; however, anti-VEGFA treatment prevented it without fully suppressing the transcriptional and metabolic programs. This study shows incongruence between single-cell transcriptional states, vascular phenotypes and related pathophysiology. Our findings also suggest that the vascular structure abnormalization, rather than neoplasms, causes the reported anti-Dll4 antibody toxicity.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
NSC23766 trihydrochloride, ≥97% (HPLC)