Merck
  • The tumor suppressor adenomatous polyposis coli regulates T lymphocyte migration.
The tumor suppressor adenomatous polyposis coli regulates T lymphocyte migration.
Science advances (2022-04-14)
Marta Mastrogiovanni, Pablo Vargas, Thierry Rose, Céline Cuche, Elric Esposito, Marie Juzans, Hélène Laude, Amandine Schneider, Mathilde Bernard, Sophie Goyard, Charlotte Renaudat, Marie-Noëlle Ungeheuer, Jérôme Delon, Andrés Alcover, Vincenzo Di Bartolo
ABSTRACT
Adenomatous polyposis coli (APC) is a tumor suppressor whose mutations underlie familial adenomatous polyposis (FAP) and colorectal cancer. Although its role in intestinal epithelial cells is well characterized, APC importance in T cell biology is ill defined. APC regulates cytoskeleton organization, cell polarity, and migration in various cell types. Here, we address whether APC plays a role in T lymphocyte migration. Using a series of cell biology tools, we unveiled that T cells from FAP patients carrying APC mutations display impaired adhesion and motility in constrained environments. We further dissected the cellular mechanisms underpinning these defects in APC-depleted CEM T cell line that recapitulate the phenotype observed in FAP T cells. We found that APC affects T cell motility by modulating integrin-dependent adhesion and cytoskeleton reorganization. Hence, APC mutations in FAP patients not only drive intestinal neoplasms but also impair T cell migration, potentially contributing to inefficient antitumor immunity.
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-Tubulin Antibody, beta, clone KMX-1, clone KMX-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1, activated, clone HUTS-4, Alexa Fluor 647 Conjugate, clone HUTS-4, from mouse, ALEXA FLUOR 647
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder