Merck
  • Resolvin D1 prevents cadmium chloride-induced memory loss and hippocampal damage in rats by activation/upregulation of PTEN-induced suppression of PI3K/Akt/mTOR signaling pathway.
Resolvin D1 prevents cadmium chloride-induced memory loss and hippocampal damage in rats by activation/upregulation of PTEN-induced suppression of PI3K/Akt/mTOR signaling pathway.
Clinical and experimental pharmacology & physiology (2021-09-28)
Ali A Shati, Attalla F El-Kott, Mahmoud A Alkhateeb
ABSTRACT
This study evaluated the protective effect of resolvin D1 (RVD1) against cadmium chloride (CdCl2 )-induced hippocampal damage and memory loss in rats and investigated if such protection is mediated by modulating the PTEN/PI3K/Akt/mTOR pathway. Adult male Wistar rats (n = 18/group) were divided as control, control + RVD1, CdCl2 , CdCl2  + RVD1 and CdCl2  + RVD1 + bpV(pic), a PTEN inhibitor. All treatments were conducted for 4 weeks. Resolvin D1 improved the memory function as measured by Morris water maze (MWM), preserved the structure of CA1 area of the hippocampus, and increased hippocampal levels of RVD1 in the CdCl2 -treated rats. Resolvin D1 also suppressed the generation of reactive oxygen species (ROS), tumour necrosis factor-α and interleukine-6 (IL-6), inhibited nuclear factor κB (NF-κB) p65, stimulated levels of glutathione (GSH), manganese superoxide dismutase (MnSOD), and Bcl2 but reduced the expression of Bax and cleaved caspase 3 in hippocampi of CdCl2 -treated rats. Concomitantly, it stimulated levels and activity of PTEN and reduced the phosphorylation (activation) of PI3K, Akt and mTOR in hippocampi of CdCl2 -treated rats. In conclusion, RVD1 attenuates CdCl2 -induced memory loss and hippocampal damage in rats mainly by activating PTEN-induced suppression of PI3K/Akt/mTOR, an effect that seems secondary to its' anti-oxidant and anti-inflammatory potential.
Sigma-Aldrich
bpV(pic), ≥95% V basis
Sigma-Aldrich
Ketamine hydrochloride/xylazine hydrochloride solution