Merck
  • Sulbactam and Colistin Susceptibility Pattern Among Multidrug-Resistant Acinetobacter Isolates From Respiratory Samples.
Sulbactam and Colistin Susceptibility Pattern Among Multidrug-Resistant Acinetobacter Isolates From Respiratory Samples.
Cureus (2022-03-08)
Mallika Sengupta, Sayantan Banerjee
ABSTRACT
Background Acinetobacter species are known to be important hospital-acquired pathogens. Unfortunately, multidrug-resistant Acinetobacter spp. has very limited options for an effective treatment. Aim To identify the common pathogens causing lower respiratory tract infections (LRTI), their antimicrobial susceptibility pattern, and determine the minimum inhibitory concentration (MIC) of sulbactam and colistin for Acinetobacter spp. Materials and methods A prospective study was done for a period of six months in a tertiary care hospital in Eastern India. The organisms causing LRTI were identified by conventional biochemical techniques and VITEK 2 Compact System (bioMérieux Inc., France). Antimicrobial susceptibility testing was performed using the Kirby‑Bauer disc diffusion method. MIC was also measured for Acinetobacter spp. to confirm certain antimicrobial agents using E-strips and micro broth dilution techniques. Results A total of 542 sputum and endotracheal tube aspirate (ETA) samples were examined during the study period. Among these, 109 samples showed growth of significant colony count of one or two organisms, yielding a sum of 115 isolates. Among these, there were 51 (44.35%) isolates of Klebsiella pneumoniae, 32 (27.83%) isolates of Pseudomonas spp., 30 (26.09%) isolates of Acinetobacter spp., and two (1.74%) isolates of Stenotrophomonas maltophilia. Although they were susceptible to colistin, Acinetobacter spp. was highly resistant to sulbactam. Conclusion Although colistin susceptibility was noted, the common pathogens causing LRTI were highly resistant to most drugs. Therefore, the causative organisms of LRTI and their susceptibility pattern should be determined to manage these cases effectively.
Sigma-Aldrich
Sulbactam
Pricing and availability is not currently available.