Merck
  • CNTF protects oligodendrocytes from ammonia toxicity: intracellular signaling pathways involved.
CNTF protects oligodendrocytes from ammonia toxicity: intracellular signaling pathways involved.
Neurobiology of disease (2008-11-11)
Laurène Cagnon, Olivier Braissant
ABSTRACT
In pediatric patients, hyperammonemia can provoke irreversible damages to developing CNS like cortical atrophy, ventricular enlargement, demyelination or gray and white matter hypodensities which are concordant with alterations of neurons and oligodendrocytes. Cerebral injury triggers endogenous protective mechanisms that can prevent or limit brain damage. Understanding these mechanisms may lead to new therapeutic strategies. We investigated whether ciliary neurotrophic factor (CNTF), a cytokine-like protein expressed by astrocytes and described as an injury-associated survival factor, was up-regulated by ammonia in developing reaggregated 3D brain cell cultures. We showed that CNTF is up-regulated by ammonia exposure, through mediation of p38 MAPK activation in astrocytes. We also observed that SAPK/JNK and Erk1/2 activations in oligodendrocytes and neurons, respectively, also play indirect roles in CNTF synthesis by astrocytes. Co-treatment with exogenous CNTF demonstrated strong protective effects on oligodendrocytes, but not on neurons, against ammonia toxicity. These protective effects involved JAK/STAT, SAPK/JNK and c-jun proteins.
Sigma-Aldrich
Anti-Ciliary Neurotrophic Factor Antibody, clone 4-68, clone 4-68, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®