Merck
  • An optimized protocol for PD-L1 pathological assessment with patient sample deglycosylation to improve correlation with therapeutic response.
An optimized protocol for PD-L1 pathological assessment with patient sample deglycosylation to improve correlation with therapeutic response.
STAR protocols (2022-03-05)
Ying-Nai Wang, Heng-Huan Lee, Yongkun Wei, Yu-Yi Chu, Weiya Xia, Michael Wang, Dihua Yu, Mien-Chie Hung
ABSTRACT
Immunotherapy via PD-1/PD-L1 blockade is a promising strategy to eradicate cancer cells. However, the PD-L1 pathological level is inconsistent with the therapeutic response and is not a reliable biomarker to stratify patients for anti-PD-1/PD-L1 therapy. Here, we describe patient sample deglycosylation in an immunohistochemistry (IHC) assay to resolve this challenge. This protocol facilitates antigen retrieval by removing N-glycans from surface antigens on formalin-fixed paraffin-embedded (FFPE) tissue slides and can be applied in medical pathology for multiple cancer types. For complete details on the use and execution of this profile, please refer to Lee et al. (2019).
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Pricing and availability is not currently available.