Merck
A phase 1 study investigating DX-2930 in healthy subjects.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology (2014-07-02)
Yung Chyung, Bradley Vince, Ryan Iarrobino, Dan Sexton, Jon Kenniston, Ryan Faucette, Chris TenHoor, Leslie E Stolz, Chris Stevens, Joseph Biedenkapp, Burt Adelman
ABSTRACT
DX-2930 is a human monoclonal antibody inhibitor of plasma kallikrein under investigation for long-term prophylaxis of hereditary angioedema. To assess the safety, tolerability, pharmacokinetics, and pharmacodynamics of DX-2930 in healthy subjects. A single-center, double-blinded study was performed in 32 healthy subjects randomized 3:1 to receive a single subcutaneous administration of DX-2930 or placebo within 1 of 4 sequential, ascending dose cohorts (n = 8 each): 0.1, 0.3, 1.0, or 3.0 mg/kg. No dose-limiting toxicity was observed. Headache was the most commonly reported treatment emergent adverse event (AE), occurring at a rate of 25% in the DX-2930- and placebo-treated groups; none were severe and all resolved. There were no serious AEs, discontinuations owing to an AE, or deaths. Two subjects had a severe AE reported as related to treatment by the blinded investigator; the 2 AEs were asymptomatic creatinine phosphokinase elevations of 902 U/L in 1 subject receiving 0.1 mg/kg DX-2930 and 1,967 U/L in 1 subject receiving placebo. For the 0.1-, 0.3-, 1.0-, and 3.0-mg/kg dose groups, respectively, mean maximum plasma concentrations were 0.6, 1.4, 5.6, and 14.5 μg/mL and mean elimination half-lives were 20.6, 16.8, 17.6, and 21.2 days. Exploratory biomarker assays, involving ex vivo activation of the kallikrein pathway, showed dose- and time-dependent inhibition of plasma kallikrein, with evidence of sustained bioactivity consistent with the pharmacokinetics profile. A single administration of DX-2930 in healthy subjects up to doses of 3.0 mg/kg was well tolerated without dose-limiting toxicity. Pharmacokinetic and pharmacodynamic data provide evidence for a long-acting biological effect relevant to long-term prophylaxis for hereditary angioedema with C1-inhibitor deficiency. ClinicalTrials.gov identifier: NCT01923207.
Sigma-Aldrich
Pro-Phe-Arg-7-amido-4-methylcoumarin, ≥95%