Merck
  • The PP2A-Integrator-CDK9 axis fine-tunes transcription and can be targeted therapeutically in cancer.
The PP2A-Integrator-CDK9 axis fine-tunes transcription and can be targeted therapeutically in cancer.
Cell (2021-05-19)
Stephin J Vervoort, Sarah A Welsh, Jennifer R Devlin, Elisa Barbieri, Deborah A Knight, Sarah Offley, Stefan Bjelosevic, Matteo Costacurta, Izabela Todorovski, Conor J Kearney, Jarrod J Sandow, Zheng Fan, Benjamin Blyth, Victoria McLeod, Joseph H A Vissers, Karolina Pavic, Ben P Martin, Gareth Gregory, Elena Demosthenous, Magnus Zethoven, Isabella Y Kong, Edwin D Hawkins, Simon J Hogg, Madison J Kelly, Andrea Newbold, Kaylene J Simpson, Otto Kauko, Kieran F Harvey, Michael Ohlmeyer, Jukka Westermarck, Nathanael Gray, Alessandro Gardini, Ricky W Johnstone
ABSTRACT
Gene expression by RNA polymerase II (RNAPII) is tightly controlled by cyclin-dependent kinases (CDKs) at discrete checkpoints during the transcription cycle. The pausing checkpoint following transcription initiation is primarily controlled by CDK9. We discovered that CDK9-mediated, RNAPII-driven transcription is functionally opposed by a protein phosphatase 2A (PP2A) complex that is recruited to transcription sites by the Integrator complex subunit INTS6. PP2A dynamically antagonizes phosphorylation of key CDK9 substrates including DSIF and RNAPII-CTD. Loss of INTS6 results in resistance to tumor cell death mediated by CDK9 inhibition, decreased turnover of CDK9 phospho-substrates, and amplification of acute oncogenic transcriptional responses. Pharmacological PP2A activation synergizes with CDK9 inhibition to kill both leukemic and solid tumor cells, providing therapeutic benefit in vivo. These data demonstrate that fine control of gene expression relies on the balance between kinase and phosphatase activity throughout the transcription cycle, a process dysregulated in cancer that can be exploited therapeutically.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-GST Tag Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12, culture supernatant, clone 4E12, from rat
Sigma-Aldrich
Anti-INTS1 Antibody, clone 4.47, clone 4.47, from mouse
Sigma-Aldrich
Anti-INTS8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-RNA polymerase II Antibody, clone CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2), clone 3E10 (rat monoclonal)., culture supernatant, clone 3E10, from rat