Merck
  • Skeletal muscle derived Musclin protects the heart during pathological overload.
Skeletal muscle derived Musclin protects the heart during pathological overload.
Nature communications (2022-01-12)
Malgorzata Szaroszyk, Badder Kattih, Abel Martin-Garrido, Felix A Trogisch, Gesine M Dittrich, Andrea Grund, Aya Abouissa, Katja Derlin, Martin Meier, Tim Holler, Mortimer Korf-Klingebiel, Katharina Völker, Tania Garfias Macedo, Cristina Pablo Tortola, Michael Boschmann, Nora Huang, Natali Froese, Carolin Zwadlo, Mona Malek Mohammadi, Xiaojing Luo, Michael Wagner, Julio Cordero, Robert Geffers, Sandor Batkai, Thomas Thum, Nadja Bork, Viacheslav O Nikolaev, Oliver J Müller, Hugo A Katus, Ali El-Armouche, Theresia Kraft, Jochen Springer, Gergana Dobreva, Kai C Wollert, Jens Fielitz, Stephan von Haehling, Michaela Kuhn, Johann Bauersachs, Joerg Heineke
ABSTRACT
Cachexia is associated with poor prognosis in chronic heart failure patients, but the underlying mechanisms of cachexia triggered disease progression remain poorly understood. Here, we investigate whether the dysregulation of myokine expression from wasting skeletal muscle exaggerates heart failure. RNA sequencing from wasting skeletal muscles of mice with heart failure reveals a reduced expression of Ostn, which encodes the secreted myokine Musclin, previously implicated in the enhancement of natriuretic peptide signaling. By generating skeletal muscle specific Ostn knock-out and overexpressing mice, we demonstrate that reduced skeletal muscle Musclin levels exaggerate, while its overexpression in muscle attenuates cardiac dysfunction and myocardial fibrosis during pressure overload. Mechanistically, Musclin enhances the abundance of C-type natriuretic peptide (CNP), thereby promoting cardiomyocyte contractility through protein kinase A and inhibiting fibroblast activation through protein kinase G signaling. Because we also find reduced OSTN expression in skeletal muscle of heart failure patients, augmentation of Musclin might serve as therapeutic strategy.
Sigma-Aldrich
PKG Iα Inhibitor, Cell-Permeable, The PKG Iα Inhibitor, Cell-Permeable controls the biological activity of PKG Iα. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Millipore
Phosphatase Inhibitor Cocktail Set V, 50X, Phosphatase Inhibitor Cocktail Set V is a 50X cocktail of four serine/threonine and tyrosine phosphatase inhibitors.
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution