Merck
  • Bone morphogenetic protein 8B promotes the progression of non-alcoholic steatohepatitis.
Bone morphogenetic protein 8B promotes the progression of non-alcoholic steatohepatitis.
Nature metabolism (2020-07-23)
Michele Vacca, Jack Leslie, Samuel Virtue, Brian Y H Lam, Olivier Govaere, Dina Tiniakos, Sophie Snow, Susan Davies, Kasparas Petkevicius, Zhen Tong, Vivian Peirce, Mette Juul Nielsen, Zsuzsanna Ament, Wei Li, Tomasz Kostrzewski, Diana Julie Leeming, Vlad Ratziu, Michael E D Allison, Quentin M Anstee, Julian L Griffin, Fiona Oakley, Antonio Vidal-Puig
ABSTRACT
Non-alcoholic steatohepatitis (NASH) is characterized by lipotoxicity, inflammation and fibrosis, ultimately leading to end-stage liver disease. The molecular mechanisms promoting NASH are poorly understood, and treatment options are limited. Here, we demonstrate that hepatic expression of bone morphogenetic protein 8B (BMP8B), a member of the transforming growth factor beta (TGFβ)-BMP superfamily, increases proportionally to disease stage in people and animal models with NASH. BMP8B signals via both SMAD2/3 and SMAD1/5/9 branches of the TGFβ-BMP pathway in hepatic stellate cells (HSCs), promoting their proinflammatory phenotype. In vivo, the absence of BMP8B prevents HSC activation, reduces inflammation and affects the wound-healing responses, thereby limiting NASH progression. Evidence is featured in primary human 3D microtissues modelling NASH, when challenged with recombinant BMP8. Our data show that BMP8B is a major contributor to NASH progression. Owing to the near absence of BMP8B in healthy livers, inhibition of BMP8B may represent a promising new therapeutic avenue for NASH treatment.
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)
Sigma-Aldrich
L-15 Medium (Leibovitz), With L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 568 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 633 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody