Merck
  • High glucose induces Nox4 expression and podocyte apoptosis through the Smad3/ezrin/PKA pathway.
High glucose induces Nox4 expression and podocyte apoptosis through the Smad3/ezrin/PKA pathway.
Biology open (2020-10-14)
Wanxu Guo, Hang Gao, Wei Pan, Panapn Yu, Guanghua Che
ABSTRACT
Podocytes are the major target in proteinuric kidney diseases such as diabetic nephropathy. The underlying molecular mechanisms by which high glucose (HG) results in podocyte damage remain unclear. This study investigated the regulatory role of Smad3, ezrin, and protein kinase A (PKA) in NADPH oxidase (Nox4) expression, reactive oxidative species (ROS) production, and apoptosis in HG-treated podocytes. A human podocyte cell line was cultured and differentiated, then treated with 30 mM HG. Apoptosis and intracellular ROS levels were assessed using TUNEL and DCF assays, respectively. Expressions of Nox4, phospho-Smad3Ser423/425, phospho-PKAThr197, and phospho-ezrinThr567 were evaluated using western blotting. ELISA was used to quantify intracellular cAMP concentration and PKA activity. Knockdown assay was used to inhibit the expressions of Smad3, Nox4, and ezrin by lentiviral shRNA. In HG-treated podocytes, the level of phospho-Smad3Ser423/425 and phospho-ezrinThr567 was increased significantly, which was accompanied by the reduction of cAMP and phospho-PKAThr197. HG-induced apoptosis was significantly prevented by the Smad3-inhibitor SIS3 or shRNA-Smad3. In podocytes expressing shRNA-ezrin or shRNA-Nox4, apoptosis was remarkably mitigated following HG treatment. HG-induced upregulation of phospho-ezrinThr567 and downregulation of phospho-PKAThr197 was significantly prevented by SIS3, shRNA-ezrin or shRNA-Smad3. Forskolin, a PKA activator, significantly inhibited HG-mediated upregulation of Nox4 expression, ROS generation, and apoptosis. Additionally, an increase in the ROS level was prohibited in HG-treated podocytes with the knockdown of Nox4, Smad3, or ezrin. Taken together, our findings provided evidence that Smad3-mediated ezrin activation upregulates Nox4 expression and ROS production, by suppressing PKA activity, which may at least in part contribute to HG-induced podocyte apoptosis.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Ezrin Inhibitor, NSC668394, The Ezrin Inhibitor, NSC668394 controls the biological activity of Ezrin.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PKA Antibody, NT, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-PKA α/β CAT (pThr197) antibody produced in rabbit, affinity isolated antibody