- Regucalcin enhances adipocyte differentiation and attenuates inflammation in 3T3-L1 cells.
Regucalcin enhances adipocyte differentiation and attenuates inflammation in 3T3-L1 cells.
Dysregulation of adipocyte differentiation and dysfunction play key roles in the pathogenesis of obesity and associated disorders such as diabetes and metabolic syndrome, and as such, a better understanding of the molecular mechanism of adipogenesis may help to elucidate the pathological condition of obesity and its associated disorders. Regucalcin (RGN) plays multiple regulatory roles in intracellular Ca2+ signaling pathways in mammalian cells. Here, we report that overexpression of RGN enhances lipid accumulation in 3T3-L1 adipocyte cells after adipogenic stimulation, accompanied by upregulation of adipocyte differentiation marker proteins. In contrast, genetic disruption of RGN inhibited adipogenic stimulation-induced differentiation of 3T3-L1 cells. Furthermore, RGN overexpression in differentiated 3T3-L1 adipocytes blocked inflammatory crosstalk between 3T3-L1 adipocytes and RAW264.7 macrophages in a transwell coculture system. Knockdown of RGN expression in cocultured 3T3-L1 adipocytes enhanced their susceptibility to RAW264.7 macrophage-mediated inflammation. These results suggest that RGN is required for 3T3-L1 adipocyte differentiation and that it exerts anti-inflammatory activity against 3T3-L1 adipocyte inflammation after coculture with RAW264.7 macrophages. Thus, RGN may be a novel regulator of adipocyte differentiation and act as a suppressor of inflammation in macrophage-infiltrated adipocyte tissue.