Merck
  • New insights into prediction of weak π-π complex association through proton-nuclear magnetic resonance analysis.
New insights into prediction of weak π-π complex association through proton-nuclear magnetic resonance analysis.
BMC chemistry (2020-12-10)
Chenyu Lin, Joseph Skufca, Richard E Partch
ABSTRACT
For analysis of weak π-π complexes proton-nuclear magnetic resonance (proton-NMR) simultaneously provides information of stacking configurations and association constants [Formula: see text] However, an apparent issue for this approach is inconsistent/impossible constant estimation which often leads to unreasonable interpretation for π-π complexation. Whether or not this proton-dependent constant variation could be attributed to simple experimental uncertainties or to more sophisticated additional unspecific shielding effects (AUS effects) was addressed by means of hypothesis tests using a robust bootstrap technique in this report. Our analysis shows the significance of AUS effects on such variation in constant estimation. A following study using numeric simulation further reveals the variation patterns induced by AUS effects and concludes that the largest [Formula: see text] among the obtained [Formula: see text] estimates of a complex is considered as the best estimate of [Formula: see text] due to minimum deviation from the true value of K and the multiple [Formula: see text] estimates of a π-π complex could provide preferable inferences for complex geometries.
Sigma-Aldrich
Terephthalaldehyde, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,3-Bis(trifluoromethyl)benzene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,3-Dinitrobenzene, 97% anhydrous basis
Pricing and availability is not currently available.