Merck
  • A Fast and Simple Method for the Determination of TBA in 18F-Labeled Radiopharmaceuticals.
A Fast and Simple Method for the Determination of TBA in 18F-Labeled Radiopharmaceuticals.
Pharmaceuticals (Basel, Switzerland) (2020-02-15)
Nils Erik Halvorsen, Ole Heine Kvernenes
ABSTRACT
A simple color spot test for determining the presence of residual tetrabutylammonium (TBA) in various 18F-radiopharmaceuticals is described. The test can be performed in less than five minutes. Iodoplatinate-saturated TLC plates are initially spotted with the 18F-radiopharmaceutical to be tested and TBA, then with deionized water or hydrogen peroxide (H2O2) solution, depending on if an antioxidant stabilizer is part of the pharmaceutical matrix. A distinct brown spot is visible at TBA concentrations of 50 µg/mL and up.
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide 30-hydrate, ≥98.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide 30-hydrate, ≥99.0% (T)