Merck
  • MANF delivery improves retinal homeostasis and cell replacement therapies in ageing mice.
MANF delivery improves retinal homeostasis and cell replacement therapies in ageing mice.
Experimental gerontology (2020-03-03)
Joana Neves, Kathleen R Chirco, Wendy Cedron-Craft, Shereen Chew, Jie Zhu, Heinrich Jasper, Deepak A Lamba
ABSTRACT
Ageing is a major risk factor for vision loss, and inflammation is an important contributor to retinal disease in the elderly. Regenerative medicine based on cell replacement strategies has emerged in recent years as a promising approach to restore vision. However, how the ageing process affects retinal homeostasis and inflammation in the retina and how this may impose a limitation to the success of such interventions remains unknown. Here we report that, in mice and humans, retinal ageing is associated with a reduction in MANF protein levels, specifically in the choroid, where increased densities of activated macrophages can be detected. We further show that the retina of old wild type mice, in the absence of any other genetic alteration, has limited homeostatic capacity after damage imposed by light exposure and reduced engraftment efficiency of exogenously supplied photoreceptors. Finally, we show that supplementation of MANF recombinant protein can improve retinal homeostasis and repair capacity in both settings, correlating with reduced numbers of activated macrophages in the old retina. Our work identifies age-related alterations in retinal homeostasis, independent of genetic alterations, leading to age-related retinal inflammation and damage susceptibility. We suggest that MANF therapy is a potential intervention to maintain retinal homeostasis in the elderly and improve the success of retinal regenerative therapies applied to aged individuals.
Sigma-Aldrich
Anti-MANF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Roche
DAPI, 4′,6-Diamidine-2′-phenylindole dihydrochloride
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%