Merck
  • Calmodulin and calmodulin-dependent protein kinase II inhibit hormone secretion in human parathyroid adenoma.
Calmodulin and calmodulin-dependent protein kinase II inhibit hormone secretion in human parathyroid adenoma.
The Journal of endocrinology (2010-10-27)
Ming Lu, Erik Berglund, Catharina Larsson, Anders Höög, Lars-Ove Farnebo, Robert Bränström
ABSTRACT
Intracellular calcium ([Ca(2+)](i)) is the most relevant modulator of parathyroid hormone (PTH) secretion. Uniquely, an increase in [Ca(2+)](i) results in an inhibition of PTH secretion, and it probably exerts its function via calcium-binding protein pathways. The ubiquitous calcium-binding proteins, calmodulin and calmodulin-dependent protein kinase II (CaMKII), have well-established roles in regulated exocytosis in neurons and neuroendocrine cells. However, their roles in parathyroid cells and PTH secretion are still unclear. Using reverse transcription-PCR and western blot analysis, we have demonstrated the expression of calmodulin and CaMKII in human normal parathyroid and parathyroid chief cell adenomas. Blocking of calmodulin and CaMKII activity by the specific antagonists calmidazolium and KN-62 respectively caused a rise in PTH secretion from parathyroid adenoma cells in spite of increased [Ca(2+)](i). The inhibitory effect of Ca(2+) calmodulin on PTH secretion may be due to the absence of synaptotagmin 1 protein in parathyroid adenomas, as demonstrated by western blot analysis. An increased extracellular calcium level acutely lowered the amount of active phosphorylated CaMKII (pCaMKII) in adenoma cells in vitro, indicating the physiological importance of this pathway. Moreover, a negative correlation between the levels of pCaMKII in parathyroid adenomas and serum calcium was found in 20 patients with primary hyperparathyroidism. Taken together, these results show that calmodulin negatively contributes to the regulation of PTH secretion in parathyroid adenoma, at least partially via a CaMKII pathway.
Sigma-Aldrich
KN-62, A cell-permeable, reversible, and selective inhibitor of CaM kinase II (Ki = 900 nM for rat brain CaM kinase II) that binds directly to the calmodulin binding site of the enzyme.
Sigma-Aldrich
KN-62, ≥95%, powder
Sigma-Aldrich
Calmidazolium chloride, solid