Merck
  • Discovery and Characterization of a Cellular Potent Positive Allosteric Modulator of the Polycomb Repressive Complex 1 Chromodomain, CBX7.
Discovery and Characterization of a Cellular Potent Positive Allosteric Modulator of the Polycomb Repressive Complex 1 Chromodomain, CBX7.
Cell chemical biology (2019-08-20)
Kelsey N Lamb, Daniel Bsteh, Sarah N Dishman, Hagar F Moussa, Huitao Fan, Jacob I Stuckey, Jacqueline L Norris, Stephanie H Cholensky, Dongxu Li, Jingkui Wang, Cari Sagum, Benjamin Z Stanton, Mark T Bedford, Kenneth H Pearce, Terry P Kenakin, Dmitri B Kireev, Gang Greg Wang, Lindsey I James, Oliver Bell, Stephen V Frye
ABSTRACT
Polycomb-directed repression of gene expression is frequently misregulated in human diseases. A quantitative and target-specific cellular assay was utilized to discover the first potent positive allosteric modulator (PAM) peptidomimetic, UNC4976, of nucleic acid binding by CBX7, a chromodomain methyl-lysine reader of Polycomb repressive complex 1. The PAM activity of UNC4976 resulted in enhanced efficacy across three orthogonal cellular assays by simultaneously antagonizing H3K27me3-specific recruitment of CBX7 to target genes while increasing non-specific binding to DNA and RNA. PAM activity thereby reequilibrates PRC1 away from H3K27me3 target regions. Together, our discovery and characterization of UNC4976 not only revealed the most cellularly potent PRC1-specific chemical probe to date, but also uncovers a potential mechanism of Polycomb regulation with implications for non-histone lysine methylated interaction partners.
Product Description
Millipore
Benzonase® Nuclease, Purity > 90%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
UNC4976 Trifluoroacetate, ≥95% (HPLC)