Merck
  • Single-Cell Survey of Human Lymphatics Unveils Marked Endothelial Cell Heterogeneity and Mechanisms of Homing for Neutrophils.
Single-Cell Survey of Human Lymphatics Unveils Marked Endothelial Cell Heterogeneity and Mechanisms of Homing for Neutrophils.
Immunity (2019-08-14)
Akira Takeda, Maija Hollmén, Denis Dermadi, Junliang Pan, Kevin Francis Brulois, Riina Kaukonen, Tapio Lönnberg, Pia Boström, Ilkka Koskivuo, Heikki Irjala, Masayuki Miyasaka, Marko Salmi, Eugene C Butcher, Sirpa Jalkanen
ABSTRACT
Lymphatic vessels form a critical component in the regulation of human health and disease. While their functional significance is increasingly being recognized, the comprehensive heterogeneity of lymphatics remains uncharacterized. Here, we report the profiling of 33,000 lymphatic endothelial cells (LECs) in human lymph nodes (LNs) by single-cell RNA sequencing. Unbiased clustering revealed six major types of human LECs. LECs lining the subcapsular sinus (SCS) of LNs abundantly expressed neutrophil chemoattractants, whereas LECs lining the medullary sinus (MS) expressed a C-type lectin CD209. Binding of a carbohydrate Lewis X (CD15) to CD209 mediated neutrophil binding to the MS. The neutrophil-selective homing by MS LECs may retain neutrophils in the LN medulla and allow lymph-borne pathogens to clear, preventing their spread through LNs in humans. Our study provides a comprehensive characterization of LEC heterogeneity and unveils a previously undefined role for medullary LECs in human immunity.
Sigma-Aldrich
Monoclonal Anti-CD15 antibody produced in mouse, clone MEM-158, purified immunoglobulin, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mannan from Saccharomyces cerevisiae, prepared by alkaline extraction
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-CLDN11 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-MARCO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)