Merck
  • Early lineage segregation of multipotent embryonic mammary gland progenitors.
Early lineage segregation of multipotent embryonic mammary gland progenitors.
Nature cell biology (2018-05-23)
Aline Wuidart, Alejandro Sifrim, Marco Fioramonti, Shigeru Matsumura, Audrey Brisebarre, Daniel Brown, Alessia Centonze, Anne Dannau, Christine Dubois, Alexandra Van Keymeulen, Thierry Voet, Cédric Blanpain
ABSTRACT
The mammary gland is composed of basal cells and luminal cells. It is generally believed that the mammary gland arises from embryonic multipotent progenitors, but it remains unclear when lineage restriction occurs and what mechanisms are responsible for the switch from multipotency to unipotency during its morphogenesis. Here, we perform multicolour lineage tracing and assess the fate of single progenitors, and demonstrate the existence of a developmental switch from multipotency to unipotency during embryonic mammary gland development. Molecular profiling and single cell RNA-seq revealed that embryonic multipotent progenitors express a unique hybrid basal and luminal signature and the factors associated with the different lineages. Sustained p63 expression in embryonic multipotent progenitors promotes unipotent basal cell fate and was sufficient to reprogram adult luminal cells into basal cells by promoting an intermediate hybrid multipotent-like state. Altogether, this study identifies the timing and the mechanisms mediating early lineage segregation of multipotent progenitors during mammary gland development.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid