
Vinpocetine Related Compound A

Synonym(s):
Ethyl (12S,13aS,13bS)-13a-ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]-naphthyridine-12-carboxylate, Ethyl vincaminate
Empirical Formula (Hill Notation):
C22H28N2O3
CAS Number:
Molecular Weight:
368.47
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2