
Verapamil Related Compound A

Synonym(s):
3,4-Dimethoxy-α-[3-(methylamino)propyl]-α-(1-methylethyl)benzeneacetonitrile monohydrochloride
Empirical Formula (Hill Notation):
C17H26N2O2 · HCl
CAS Number:
Molecular Weight:
326.86
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2