
Tyloxapol

Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol polymer with formaldehyde and oxirane
CAS Number:
Products
Product Category
Brand
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing